Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates

Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates

384-well microarray plate with pyramidal well bottoms for maximum sample recovery.
Have Questions?
Catalog NumberColorVolume (Metric) WellWell ShapeFit for Purpose
AB1056
also known as AB-1056
Natural55 μLRound, Conical Bottom (V-well)PCR Ready
Catalog number AB1056
also known as AB-1056
Price (INR)
35,806.00
Case of 50
Color:
Natural
Volume (Metric) Well:
55 μL
Well Shape:
Round, Conical Bottom (V-well)
Fit for Purpose:
PCR Ready
Price (INR)
35,806.00
Case of 50

Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

 

Highlights

  • Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
  • Pyramidal well bottom for ease of sample retrieval
  • Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination

Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Dnase FreeYes
Description384-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Max. RCF5000 x g
No. per Case50
No. per Pack10
Packaging10 pk., 50 cs.
Plate SizeANSI/ SLAS Standard
Product Type384-well plate
RNase FreeYes
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
Volume (Metric) Working45 μL
Array16 x 24
BarcodeCustom Barcoding Available
ColorNatural
Fit for PurposePCR Ready
FormatStandard
MaterialPolypropylene
No. of Wells384
Product LineAbgene
SterilityCleanroom
Surface TreatmentNon-treated
Volume (Metric) Well55 μL
Well ShapeRound, Conical Bottom (V-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

What 96-well plates do you recommend for -80 degrees C storage?

Both polypropylene and polystyrene plates can be used to store samples at -80 degrees C and depends on the downstream application. Please note that the sealing method selected is critical for storing at these temperatures and heat sealing is recommended.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What is the recommended storage temperature for Abgene 384 Well Polypropylene Sample Processing & Storage Plates?

We recommend storing Abgene 384 Well Polypropylene Sample Processing & Storage Plates at ambient temperature.