Armadillo™ Low-Profile PCR Strip Plate, 96 well, green
Armadillo™ Low-Profile PCR Strip Plate, 96 well, green
Armadillo™ Low-Profile PCR Strip Plate, 96 well, green
Armadillo™ Low-Profile PCR Strip Plate, 96 well, green
Armadillo™ Low-Profile PCR Strip Plate, 96 well, green
Thermo Scientific™

Armadillo™ Low-Profile PCR Strip Plate, 96 well, green

Thermo Scientific Armadillo 96-well low-profile PCR strip plates are the ultimate in high-performance PCR plastics. Combining the rigidity of aRead more
Have Questions?
Catalog NumberQuantity
AB2696G25 plates
Catalog number AB2696G
Price (INR)
18,693.00
25 plates
Quantity:
25 plates
Price (INR)
18,693.00
25 plates
Thermo Scientific Armadillo 96-well low-profile PCR strip plates are the ultimate in high-performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, Armadillo PCR strip plates provide superior thermal cycling performance under all conditions without warping. The plates can be easily and safely broken into 8-tube strips to help minimize waste.

Features of Armadillo 96-well low-profile PCR strip plates include:
Thermostable polycarbonate semi-skirted frame design for warp-resistance during thermal cycling and enhanced mechanical stability during robotic handling
Breakable into 8-tube strips to accommodate variable numbers of samples and minimize waste
Optically clear skirt aids in visualization of liquids and improves stacking capabilities in automated handling systems
ANSI:SBS footprint ensures complete compatibility with robotic systems for high-throughput applications
Thin-walled U-bottom wells for optimal heat transfer and maximum sample recovery
Black alpha-numeric lettering for easy well identification
Integrated frame and well design for improved heat sealing
Low profile design for maximum heat transfer and minimal evaporation with low-volume samples (200 μL working volume)
Available with both clear- and white-colored wells to help ensure the highest level of sensitivity for all PCR/qPCR reactions

Specifications
BreakapartYes
For Use With (Equipment)Real-Time PCR Systems or Thermal Cycler
Packaging25 Plates
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedLow Profile
Skirt StyleSemi
Volume (Metric) Working0.1 mL
Well ColorClear
BarcodeNo Barcode
ColorGreen
MaterialPolycarbonate
No. of Wells96
Product LineArmadillo
Volume (Metric) Well0.2 mL
Well ShapeRound Bottom (U-well)
Unit Size25 plates