Armadillo™ Low-Profile PCR Strip Plate, 96 well, clear, 5 plates
Armadillo™ Low-Profile PCR Strip Plate, 96 well, clear, 5 plates
Armadillo™ Low-Profile PCR Strip Plate, 96 well, clear, 5 plates
Armadillo™ Low-Profile PCR Strip Plate, 96 well, clear, 5 plates

Armadillo™ Low-Profile PCR Strip Plate, 96 well, clear, 5 plates

The Thermo Scientific 96-Well Semi-Skirted Plates have a low-profile, thin well wall design and are compatible with PCR and qPCR applications.
Have Questions?
Catalog NumberColor
AB2696SMPClear
Catalog number AB2696SMP
Price (INR)
-
Color:
Clear

Thermo Scientific Armadillo 96-well low-profile PCR strip plates are the ultimate in high-performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, Armadillo PCR strip plates provide superior thermal cycling performance under all conditions without warping. The plates can be easily and safely broken into 8-tube strips to help minimize waste. Additionally, up to 1 tube can be separated from the strip using the Thermo Scientific PCR strip cutting Tool (A35329).

The Armadillo 96-well low-profile PCR strip plates include Thermostable polycarbonate semi-skirted frame and are breakable into 8-tube strips to accommodate variable numbers of sample. Additionally, up to 1 tube can be separated from the strip using the Thermo Scientific PCR strip cutting Tool (A35329). The plates are Optically clear skirt with ANSI:SBS footprint and thin-walled U-bottom wells for optimal heat transfer. For easy identification, alpha-numeric lettering are marked on the plates.

 Features of Armadillo 96-well low-profile PCR strip plates include:

 

  • Thermostable polycarbonate semi-skirted frame design for warp-resistance during thermal cycling and enhanced mechanical stability during robotic handling
  • Breakable into 8-tube strips to accommodate variable numbers of samples and minimize waste
  • Optically clear skirt aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • ANSI:SBS footprint ensures complete compatibility with robotic systems for high-throughput applications
  • Thin-walled U-bottom wells for optimal heat transfer and maximum sample recover
  • Black alpha-numeric lettering for easy well identification
  • Integrated frame and well design for improved heat sealing
  • Low profile design for maximum heat transfer and minimal evaporation with low-volume samples (200 μL working volume)
Specifications
AutoclavableNot Autoclavable
For Use With (Equipment)QuantStudio™ 5 Real-Time PCR System, 7500 Fast Real-Time PCR System
ColorClear
Fit for PurposePCR Ready
MaterialPolycarbonate
No. of Wells96
Product LineArmadillo
SterilityNon-sterile
Well ShapeRound Bottom (U-well)
Unit SizeEach