Armadillo PCR Plate, 384-well, yellow, white wells
Thermo Scientific™

Armadillo PCR Plate, 384-well, yellow, white wells

Thermo Scientific Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frameRead more
Have Questions?
Catalog NumberQuantity
AB3384Y50 plates
Catalog number AB3384Y
Price (INR)
63,609.00
Each
Quantity:
50 plates
Price (INR)
63,609.00
Each
Thermo Scientific Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping. These 384-well plates are yellow, with white wells.

Features of Armadillo PCR plates:

Optically clear skirt—aids in visualization of liquids and improves stacking capabilities in automated handling systems
Raised rim design—improved well surface area improves seal integrity in automated heat sealing
ANSI:SBS footprint—with rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
Thermostable polycarbonate frame design—warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
Thin walled U-bottom wells—optimal heat transfer and maximum sample recovery
Indented Alphanumeric Grid reference lettering—integrated frame and well design for improved heat sealing
Low profile design—allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (30 μL working volume)
Barcoding available—all plates are available already barcoded and can be provided barcoded to customer specifications for fully-automated applications
White plates are ideal for luminescence-based assays

Applications
• PCR
• qPCR/Real-Time PCR
This product is sold under license from Eppendorf AG.
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging50 Plate
Product TypePCR Microplate
Quantity50 plates
Reaction SpeedStandard
Skirt StyleSkirted
Volume (Metric) Well40 μL
Volume (Metric) Working25 μL
Well ColorWhite
BarcodeNo Barcode
ColorYellow, White
MaterialPolycarbonate, Polypropylene
No. of Wells384
Product LineArmadillo
Unit SizeEach