InSight Accessories
Actual product may vary
InSight Accessories
InSight Accessories
InSight Accessories
InSight Accessories
Thermo Scientific™

InSight Accessories

Maximize your Thermo Scientific™ InSight with essential accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
ACC-CON-1004Mote commissioning cable assembly
ACC-BAT-1002Replacement C battery Li ion
ACC-CON-1010
ACC-CON-1011Internal 12V power cable for T3C1D2 mote (to chart recorder power)
ACC-CON-1030Terminal block kit
ACC-CON-1016InSightLink gateway power cable
INSIGHTLINK-1008InSightLink gateway (915MHz -> customer ethernet -> Cloud)
Catalog number ACC-CON-1004
Price (INR)
-
Description:
Mote commissioning cable assembly
Thermo Scientific™ InSight accessories include replacement batteries, mote cable assembly, terminal block kits, power cables and more.
  • Replacement C battery Li ion
  • Mote commissioning cable assembly
  • External PSU 110V for T3C1D2 mote
  • Terminal block kit
  • InSightLink gateway power cable
  • InSightLink gateway
  • Internal 12V power cable for T3C1D2 mote
  • Warranty varies by country, please contact us for details
Not available outside North America
Specifications
DescriptionMote commissioning cable assembly
For Use With (Application)Cold Storage
For Use With (Equipment)Mote Commissioning Cable Assembly
TypeMote Commissioning Cable Assembly
Unit SizeEach