Iodine, crystalline, 99.99+% (metals basis)
Iodine, crystalline, 99.99+% (metals basis)
Iodine, crystalline, 99.99+% (metals basis)
Alfa Aesar

Iodine, crystalline, 99.99+% (metals basis)

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF-000158-18
HSN 28012000
Price (INR)
4,168.00
Each
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Non-Volatile Residue≤0.005%
Impurity contentChlorine and Bromine (as Cl): ≤0.005%
FormCrystals
Appearance (Color)Dark grey to black to purple
Assay from Supplier's CofA≥99.99% Iodine (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only