Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)
Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)
Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)
Alfa Aesar

Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)

Form: 90% 2.5-25mm (0.1-1.0 in.) | CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number ALF-010213-A1
HSN 72039000
Price (INR)
4,113.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
4,113.00
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only