Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Alfa Aesar

Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
4 x 500 g
Catalog number ALF-010233-22
HSN 98020000
Price (INR)
4,610.00
Each
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7439-95-4
IUPAC Namemagnesium
Molecular FormulaMg
InChI KeyJLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size<45 µm : ≥ 92.0%
Assay from Suppliers CofA≥99.8%
Appearance (Color)Metallic white to grey
FormPowder
Particle Size>45 µm : ≤ 8.0%

Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only