Potassium nitrate, 99.994% (metals basis)
Potassium nitrate, 99.994% (metals basis)
Potassium nitrate, 99.994% (metals basis)
Alfa Aesar

Potassium nitrate, 99.994% (metals basis)

CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF-011008-22100 g
Catalog number ALF-011008-22
HSN 28342100
Price (INR)
23,711.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
23,711.00
Each
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.006% max.
Potassium nitrate has several applications. It is used as an oxidizer, food preservative, fertilizer, and an active ingredient of condensed aerosol fire suppression systems. It is a powerful nitrating agent in sulfuric acid medium. In addition, it also used as an electrolyte in a salt bridge and thermal storage medium in power generation systems. Ternary salts containing potassium nitrate have been reported to improve the heat storage capacity in the molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only