Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis)
Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis)
Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis)
Alfa Aesar

Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis)

For Catalysis, Electroplating, pHotography | CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALF-011051-03
HSN 28439019
Price (INR)
7,177.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
7,177.00
Each
Chemical Identifiers
CAS26023-84-7
IUPAC Namedihydrogen hexachloroplatinumtetrakis(ylium)
Molecular FormulaCl6H2Pt
InChI KeyZKOQTCCVSRSECD-UHFFFAOYSA-J
SMILES[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink, microscopy.Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride followed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts. In addition to this, it is also used as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only