Barium perchlorate trihydrate, Reagent Grade
Barium perchlorate trihydrate, Reagent Grade
Barium perchlorate trihydrate, Reagent Grade
Alfa Aesar

Barium perchlorate trihydrate, Reagent Grade

CAS: 10294-39-0 | BaCl2H6O11 | 390.264 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF-011656-A1
HSN 28299010
Price (INR)
25,726.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
25,726.00
Each
Chemical Identifiers
CAS10294-39-0
IUPAC Namebarium(2+) trihydrate diperchlorate
Molecular FormulaBaCl2H6O11
InChI KeyNBHDMFAHVJWOHD-UHFFFAOYSA-L
SMILESO.O.O.[Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ba 33.8-36.5%
Barium perchlorate, trihydrate, reagent is an extremely powerful oxidizing agent, and therefore commonly used in industry and pyrotechnics. It is also used in RPE-For analysis. It is used as an oxidizing agent, for gas drying, for ribonuclease determination and in the water analysis for sulfate titration with Thorin as indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only