Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.995% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.995% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.995% (metals basis)
Alfa Aesar

Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.995% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF-012058-FF25 x 25 mm
Catalog number ALF-012058-FF
HSN 98020000
Price (INR)
102,074.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (INR)
102,074.00
Each
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver to gray
Comment0.22-0.28mm thick
Total Metal Impurities0.005% max.
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only