Barium nitrate, ACS, 99+%
Barium nitrate, ACS, 99+%
Barium nitrate, ACS, 99+%
Alfa Aesar

Barium nitrate, ACS, 99+%

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
2 kg
Catalog number ALF-012194-36
HSN 28342930
Price (INR)
2,269.00
Each
Quantity:
500 g
Request bulk or custom format
Price (INR)
2,269.00
Each
Chemical Identifiers
CAS10022-31-8
IUPAC Namebarium(2+) dinitrate
Molecular FormulaBaN2O6
InChI KeyIWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals(ICP-OMS) 5ppm
Impurity contentSr 0.1%
pHof a 5% solution 5.0-8.0 at 25?
Assay (unspecified)99.0% Ba(NO₃)₂ min
Impurity contentFe 2ppm
View more
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only