Tellurium(IV) chloride, 99.9% (metals basis)
Tellurium(IV) chloride, 99.9% (metals basis)
Tellurium(IV) chloride, 99.9% (metals basis)
Alfa Aesar

Tellurium(IV) chloride, 99.9% (metals basis)

CAS: 10026-07-0 | Cl4Te | 269.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF-012284-14
HSN 28273990
Price (INR)
13,912.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
13,912.00
Each
Chemical Identifiers
CAS10026-07-0
IUPAC Nametetrachloro-λ⁴-tellane
Molecular FormulaCl4Te
InChI KeySWLJJEFSPJCUBD-UHFFFAOYSA-N
SMILESCl[Te](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Tellurium(IV) chloride is used in organic synthesis to prepare Cl-C-C-TeCl3derivatives by reacting with alkenes and aryl tellurium compounds by reacting with electron rich arenes. It is involved in the preparation of diaryl telluride from anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only