Aluminum hydroxide, 76.5% min
Aluminum hydroxide, 76.5% min
Aluminum hydroxide, 76.5% min
Alfa Aesar

Aluminum hydroxide, 76.5% min

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF-012366-A32 kg
Catalog number ALF-012366-A3
HSN 28183000
Price (INR)
43,695.00
Each
Quantity:
2 kg
Request bulk or custom format
Chemical Identifiers
CAS21645-51-2
IUPAC Namealuminium(3+) trihydroxide
Molecular FormulaAlH3O3
InChI KeyWNROFYMDJYEPJX-UHFFFAOYSA-K
SMILES[OH-].[OH-].[OH-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Chloride content≤0.85%
pH≤10.0
FormPowder, amorphous
Assay (special)Acid Neutralizing Capacity: ≥25.0meq/g
View more
As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmeticsAluminum hydroxide is used as a desiccant powder, filler in paper, plastics, rubber and cosmetics. It is used as a smoke suppressant and mordant dye. It is also used in drugs as antacid and antihyperphosphatemic. It is also used as a Claus catalyst support for waterproof fabrics. It is an important starting material for the preparation of other aluminum compounds, calcined alumina, aluminum sulfate, polyaluminum chloride, zeolites, sodium aluminate, activated alumina and aluminum nitrate. In addition, it is used as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only