Potassium nitrate, ACS, 99.0% min
Potassium nitrate, ACS, 99.0% min
Potassium nitrate, ACS, 99.0% min
Alfa Aesar

Potassium nitrate, ACS, 99.0% min

For Pyrotechnics, Glass Manufacture, Tempering Steel | CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number ALF-013443-A3
HSN 28342100
Price (INR)
3,892.00
Each
Quantity:
2 kg
Request bulk or custom format
Price (INR)
3,892.00
Each
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormSolid
Assay (unspecified)≥99.0% (KNO3)
Chloride content≤0.002%
Impurity contentCalcium (Ca): ≤0.005%
View more
Pyrotechnics, glass manufacture, tempering steel, reagentPotassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only