Sodium 1-heptanesulfonate monohydrate, 99%
Sodium 1-heptanesulfonate monohydrate, 99%
Sodium 1-heptanesulfonate monohydrate, 99%
Alfa Aesar

Sodium 1-heptanesulfonate monohydrate, 99%

CAS: 207300-90-1 | C7H17NaO4S | 220.259 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF-015214-18
HSN 29041090
Price (INR)
11,011.00
Each
Quantity:
50 g
Request bulk or custom format
Price (INR)
11,011.00
Each
Chemical Identifiers
CAS207300-90-1
IUPAC Namesodium heptane-1-sulfonate hydrate
Molecular FormulaC7H17NaO4S
InChI KeyXWZCREJRXRKIRQ-UHFFFAOYSA-M
SMILESO.[Na+].CCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at200nm: <0.300 AU ( 0.25M aq. solution)
Optical Absorbance at210nm: <0.200 AU ( 0.25M aq. solution)
Solution Test10% aq. solution: Clear and colorless
Sodium 1-heptanesulfonate monohydrate is an ion pairing reagent used in the HPLC analysis of proteins and peptides. It is utilized in high performance capillary electrophoresis analysis of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only