Tin(II) acetate, 95%
Tin(II) acetate, 95%
Tin(II) acetate, 95%
Alfa Aesar

Tin(II) acetate, 95%

CAS: 638-39-1 | C4H6O4Sn | 236.798 g/mol
Have Questions?
Catalog NumberQuantity
ALF-022361-065 g
Catalog number ALF-022361-06
HSN 29152990
Price (INR)
10,903.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
10,903.00
Each
Chemical Identifiers
CAS638-39-1
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Tin(II) acetate is used as a precursor in the production of tin anode, Sn-Cu bimetallic nanoparticles, high surface area tin oxide catalysts. It acts as Lewis acid catalysts for the esterification and transesterification reactions. It serves as an efficient catalytic system for carbon-hydrogen activation of methoxy group of anisole in presence of palladium(II) acetate. It plays an important role in stannous oral care compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only