Water, ultrapure, HPLC Grade
Water, ultrapure, HPLC Grade
Water, ultrapure, HPLC Grade
Alfa Aesar

Water, ultrapure, HPLC Grade

CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 4 L
Catalog number ALF-022934-K2
HSN 28530010
Price (INR)
5,659.00
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
5,659.00
Each
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at≤0.005 AU @ 400nm (non-U.S. specification)
Optical Absorbance at≤0.01 AU @ 195nm (non-U.S. specification)
Optical Absorbance at≤0.01 AU @ 200nm (non-U.S. specification)
Optical Absorbance at400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max (U.S. specification)
Residue on Evaporation≤2 ppm
View more
Water ultrapure is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is also used in conductometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only