Tricyclohexylphosphine, 96%
Tricyclohexylphosphine, 96%
Tricyclohexylphosphine, 96%
Alfa Aesar

Tricyclohexylphosphine, 96%

CAS: 2622-14-2 | C18H33P | 280.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALF-030386-14
HSN 29319090
Price (INR)
42,122.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
42,122.00
Each
Specifications
Chemical Name or MaterialTricyclohexylphosphine
CAS2622-14-2
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Tricyclohexylphosphine is used as a ligand in organometallic chemistry. As a reagent, it is used for a range of metal-catalyzed organic transformations. Crabtree's catalyst is an organoiridium compound containing tricyclohexyl phosphine used for hydrogenation of mono-, di-, tri-, and tetra- substituted substrates and hydrogen transfer reactions. Grubbs’catalysts consists of transition metal ruthenium and tricylohexyl phosphine utilized in olefin metathesis in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only