Lithium metaborate, anhydrous, tech.
Lithium metaborate, anhydrous, tech.
Lithium metaborate, anhydrous, tech.
Alfa Aesar

Lithium metaborate, anhydrous, tech.

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF-032576-22
HSN 28402090
Price (INR)
20,619.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
20,619.00
Each
Chemical Identifiers
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95% min
Lithium Metaborate is commonly used as a fused compound or a component for an assay. It has been used for the analysis of wild rice for silicon, calcium, magnesium and potassium. Lithium metaborate was used to fuse with solid samples for decomposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only