Oxalic acid, 0.1N Standardized Solution
Oxalic acid, 0.1N Standardized Solution
Oxalic acid, 0.1N Standardized Solution
Alfa Aesar

Oxalic acid, 0.1N Standardized Solution

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Catalog NumberQuantity
ALF-035622-K21 L
Catalog number ALF-035622-K2
HSN 29171110
Price (INR)
8,226.00
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
8,226.00
Each
Specifications
Chemical Name or MaterialOxalic acid
CAS144-62-7
Recommended StorageAmbient temperatures
EINECS Number205-634-3
TSCAYes
View more
Oxalic acid, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only