Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Alfa Aesar

Selenium powder, -200 mesh, 99.999% (metals basis)

CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF-036208-1850 g
Catalog number ALF-036208-18
HSN 28049000
Price (INR)
32,764.00
Each
Quantity:
50 g
Request bulk or custom format
Price (INR)
32,764.00
Each
Chemical Identifiers
CAS7782-49-2
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤ 0.001% (≤10ppm)
Particle Size≥90% is ≤74µm (≤200mesh)
Appearance (Color)Pale grey to dark grey
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only