Tungsten(V) ethoxide
Tungsten(V) ethoxide
Tungsten(V) ethoxide
Alfa Aesar

Tungsten(V) ethoxide

Intermediate for sol-gel coatings | CAS: 26143-11-3 | C10H25O5W | 409.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF-041794-09
HSN 29051990
Price (INR)
97,272.00
Each
Quantity:
10 g
Request bulk or custom format
Price (INR)
97,272.00
Each
Chemical Identifiers
CAS26143-11-3
IUPAC Namepentaethoxytungsten
Molecular FormulaC10H25O5W
InChI KeySRGJJDPFERQMIL-UHFFFAOYSA-N
SMILESCCO[W](OCC)(OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Specific Gravity1.44 to 1.47
Appearance (Color)Clear to hazy, colorless to yellow to purple
FormLiquid
Tungsten(V) ethoxide is used as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only