n-Octacosane, 99%
n-Octacosane, 99%
n-Octacosane, 99%
Alfa Aesar

n-Octacosane, 99%

CAS: 630-02-4 | C28H58 | 394.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number ALF-042460-18
HSN 29011000
Price (INR)
8,943.00
Each
Quantity:
50 g
Request bulk or custom format
Price (INR)
8,943.00
Each
Chemical Identifiers
CAS630-02-4
SpecificationsSpecification SheetSpecification Sheet
FormWaxy flake or powder
Appearance (Color)White
Assay (GC)≥ 98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only