Chromate, Ion chromatography standard solution, Specpure™ CrO4∣-2 1000μg/mL
Alfa Aesar

Chromate, Ion chromatography standard solution, Specpure™ CrO4∣-2 1000μg/mL

K2CrO4 in H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALF-042564-AE
HSN 28415090
Price (INR)
14,160.00
Each
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialChromate
Physical FormLiquid
Name NoteCrO4-2 1000μg/mL
Health Hazard 1H340-H350i
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only