Acetone, Environmental Grade, 99.5+%
Acetone, Environmental Grade, 99.5+%
Acetone, Environmental Grade, 99.5+%
Alfa Aesar

Acetone, Environmental Grade, 99.5+%

Suitable For Pesticide Residue Analysis | CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 4 L
1 L
4 L
Catalog number ALF-043053-M6
HSN 29141100
Price (INR)
68,372.00
Each
Quantity:
4 x 4 L
Request bulk or custom format
Price (INR)
68,372.00
Each
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation5ppm
Residual halide(as Heptachlorepoxide by GC/ECD) 10ng/L
Residual water0.5%
Suitable for pesticide residue analysisAcetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glassware due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon and photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only