Sulfur powder, -325 mesh, 99.5%
Sulfur powder, -325 mesh, 99.5%
Sulfur powder, -325 mesh, 99.5%
Sulfur powder, -325 mesh, 99.5%
Alfa Aesar

Sulfur powder, -325 mesh, 99.5%

CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Catalog NumberQuantity
ALF-043766-36500 g
Catalog number ALF-043766-36
HSN 28020010
Price (INR)
6,170.00
Each
Quantity:
500 g
Request bulk or custom format
Price (INR)
6,170.00
Each
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only