Amberlyst™ 15(H), wet, ion exchange resin
Amberlyst™ 15(H), wet, ion exchange resin
Amberlyst™ 15(H), wet, ion exchange resin
Alfa Aesar

Amberlyst™ 15(H), wet, ion exchange resin

CAS: 39389-20-3 | C18H18O3S | 314.399 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number ALF-044079-A1
HSN 38229090
Price (INR)
9,414.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
9,414.00
Each
Chemical Identifiers
CAS39389-20-3
IUPAC Name1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
Molecular FormulaC18H18O3S
InChI KeySIWVGXQOXWGJCI-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Opaque
Moisture holding capacity52 to 57% ( in the H form)
Exchange capacity>/= 1.7 meq/ml
FormBeads
Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only