Diaion™ SK104(Na), ion exchange resin, 4% cross-linked, strongly acidic gel type, 1.2 meq/ml on PS-DVB, Thermo Scientific™
Alfa Aesar

Diaion™ SK104(Na), ion exchange resin, 4% cross-linked, strongly acidic gel type, 1.2 meq/ml on PS-DVB, Thermo Scientific™

Specialty process and amino acid separations
Have Questions?
Catalog NumberQuantity
ALF-046452-A11 kg
Catalog number ALF-046452-A1
HSN 39140090
Price (INR)
6,396.00
Each
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialDiaion™ SK104 (Na), ion exchange resin
Physical Form300-1180μm Beads
Name Note4% cross-linked, strongly acidic gel type, 1.2meq/mL on PS-DVB
Recommended StorageAmbient temperatures
TSCANo
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.