Sodium isopropoxide
Sodium isopropoxide
Sodium isopropoxide
Alfa Aesar

Sodium isopropoxide

CAS: 683-60-3 | C3H7NaO | 82.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALF-089446-14
HSN 29051990
Price (INR)
21,889.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
21,889.00
Each
Chemical Identifiers
CAS683-60-3
IUPAC Namesodium propan-2-olate
Molecular FormulaC3H7NaO
InChI KeyWBQTXTBONIWRGK-UHFFFAOYSA-N
SMILES[Na+].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormPowder
Chloride content≤0.01%
Total Metal Impurities≤1 %
Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only