1-Ethyl-2-methylquinolinium iodide, 97%
1-Ethyl-2-methylquinolinium iodide, 97%
1-Ethyl-2-methylquinolinium iodide, 97%
1-Ethyl-2-methylquinolinium iodide, 97%
Alfa Aesar

1-Ethyl-2-methylquinolinium iodide, 97%

CAS: 606-55-3 | C12H14IN | 299.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number ALF-A10584-06
HSN 29334900
Price (INR)
4,286.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (INR)
4,286.00
Each
Add to cart
Chemical Identifiers
CAS606-55-3
IUPAC Name1-ethyl-2-methylquinolin-1-ium iodide
Molecular FormulaC12H14IN
InChI KeyOEVSHJVOKFWBJY-UHFFFAOYSA-M
SMILES[I-].CC[N+]1=C2C=CC=CC2=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormPowder
Assay (Titration ex Iodide)≥96.0 to ≤104.0%
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

Solubility
Soluble in Water.

Notes
Precursor in synthesis of carbocyanine dyes.
RUO – Research Use Only
Seiki Sakanoue; Yasushi Kai1; Noritake Yasuoka1; Nobutami Kasai1; Masao Kakudo. The Crystal Structure of 1-Ethyl-2-methylquinolinium Iodide, [(C12H14N)+·I-]. Bull. Chem. Soc. Jpn. 1970, 43, (5) 1306-1312.