Lead(II) acetate trihydrate, 99%
Lead(II) acetate trihydrate, 99%
Lead(II) acetate trihydrate, 99%
Alfa Aesar

Lead(II) acetate trihydrate, 99%

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALF-A11746-30
HSN 29152990
Price (INR)
2,499.00
Each
Quantity:
250 g
Request bulk or custom format
Price (INR)
2,499.00
Each
Chemical Identifiers
CAS6080-56-4
IUPAC Nameλ²-lead(2+) diacetate trihydrate
Molecular FormulaC4H12O7Pb
InChI KeyMCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisCalcium (Ca): ≤ 200ppm
Elemental AnalysisCopper (Cu): ≤ 200ppm
Elemental AnalysisIron (Fe): ≤ 200ppm
Elemental AnalysisSodium (Na): ≤ 200ppm
FormCrystals or crystalline powder
View more
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

Solubility
Freely soluble in water, alcohol and glycerol.

Notes
Decomposes above 200°Air and light sensitive. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Knichal, J. V.; Gee, W. J.; Burrows, A. D.; Raithby, P. R.; Wilson, C. C. A new small molecule gelator and 3D framework ligator of lead(II). CrystEngComm 2015, 17 (42), 8139-8145.
  2. Sun, X.; Yang, J.; Zhang, W.; Zhu, X.; Hu, Y.; Yang, D.; Yuan, X.; Yu, W.; Dong, J.; Wang, H.; Li, L.; Kumar, V. R.; Liang, S. Lead acetate trihydrate precursor route to synthesize novel ultrafine lead oxide from spent lead acid battery pastes. J. Power Sources 2014, 269, 565-576.