Sodium cyanate, 95%
Sodium cyanate, 95%
Sodium cyanate, 95%
Alfa Aesar

Sodium cyanate, 95%

CAS: 917-61-3 | CNNaO | 65.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number ALF-A12413-22
HSN 28429090
Price (INR)
3,452.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
3,452.00
Each
Chemical Identifiers
CAS917-61-3
IUPAC Namesodium cyanate
Molecular FormulaCNNaO
InChI KeyZVCDLGYNFYZZOK-UHFFFAOYSA-M
SMILES[Na+].[O-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (ex Sulphated Ash)≥94.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤3.5%
Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

Solubility
Soluble in water 11.6 g/100 mL (25°C). Slightly soluble in ammonia, benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. R. Badera,; D.J. Dupréa,; F. Schütz. The preparation of pure sodium cyanate from urea. Biochimica et Biophysica Acta. 1948, 2 543-548.
  2. Gavin Chit Tsui et. al. Expedient synthesis of chiral oxazolidinone scaffolds via rhodium-catalyzed asymmetric ring-opening with sodium cyanate.. Organic Letters. 2013, 15 (5),1064-1067.