Hexanoic acid, 98+%
Hexanoic acid, 98+%
Hexanoic acid, 98+%
Alfa Aesar

Hexanoic acid, 98+%

CAS: 142-62-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number ALF-A13789-AE
HSN 29159030
Price (INR)
1,793.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
1,793.00
Each
Chemical Identifiers
CAS142-62-1
IUPAC Namehexanoic acid
Molecular FormulaC6H12O2
InChI KeyFUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILESCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.0%
Appearance (Color)Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormLiquid
Identification (FTIR)Conforms
View more
Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

Solubility
Miscible with diethyl ether and ethanol. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and allyl alcohol.
RUO – Research Use Only

General References:

  1. Ramesh, S.; Cherkupally, P.; Govender, T.; Kruger, H. G.; Albericio, F.; de la Torre, B. G. 6-(Bromomaleimido)hexanoic Acid as a Connector for the Construction of Multiple Branched Peptide Platforms. Org. Lett. 2015, 17 (3), 464-467.
  2. Lee, Y.; Choi, J. W.; Suh, D. J.; Ha, J. M.; Lee, C. H. Ketonization of hexanoic acid to diesel-blendable 6-undecanone on the stable zirconia aerogel catalyst. Appl. Catal., A 2015, 506, 288-293.