1H-Pyrazole, 98%
1H-Pyrazole, 98%
1H-Pyrazole, 98%
Alfa Aesar

1H-Pyrazole, 98%

1H-Pyrazole, 98%, C3H4N2, CAS Number-288-13-1, 1-h-pyrazole, 1h-pyrazol, pyrazole, unii-3qd5kjz7zj, hpz, pyrazol, diazole, pyrazol#1, 1,2-diazole, 3qd5kjz7zj, 100g, 103775, 186 deg.C to 188 deg.C, CHEBI:17241, 206-017-1, 68.08, H302-H315-H319-H335 | CAS: 288-13-1 | C3H4N2 | 68.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF-A14186-22
HSN 29331990
Price (INR)
11,552.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
11,552.00
Each
Chemical Identifiers
CAS288-13-1
IUPAC Name1H-pyrazole
Molecular FormulaC3H4N2
InChI KeyWTKZEGDFNFYCGP-UHFFFAOYSA-N
SMILESN1C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1.0%
Assay (GC)≥97.5%
Melting Point65.0-71.0?C
Appearance (Color)White to yellow or cream
View more
1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only

General References:

  1. Michael S. Bernatowicz; Youling Wu; Gary R. Matsueda. 1H-Pyrazole-1-carboxamidine hydrochloride an attractive reagent for guanylation of amines and its application to peptide synthesis. J. Org. Chem. 1992, 57 (8), 2497-2502.
  2. Adnan A. Bekhit; Hayam M.A. Ashour; Yasser S. Abdel Ghany; Alaa El-Din A. Bekhit; Azza Baraka. Synthesis and biological evaluation of some thiazolyl and thiadiazolyl derivatives of 1H-pyrazole as anti-inflammatory antimicrobial agents. European Journal of Medicinal Chemistry. 2008, 43 (3), 456-463.
  3. Forms a complex with Pyridinium chlorochromate, A11752, which allows the selective oxidation of allylic alcohols in the presence of non-allylic alcohols: Lipids, 19, 550 (1984).
  4. Can be protected on nitrogen by hydroxymethylation with paraformaldehyde, permitting 3-lithiation with t-BuLi: Tetrahedron, 45, 4253 (1989).