4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%
Alfa Aesar

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%

CAS: 81-11-8 | C14H14N2O6S2 | 370.394 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF-A16664-22
HSN 29215990
Price (INR)
7,331.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
7,331.00
Each
Chemical Identifiers
CAS81-11-8
IUPAC Name5-amino-2-[(1E)-2-(4-amino-2-sulfophenyl)ethenyl]benzene-1-sulfonic acid
Molecular FormulaC14H14N2O6S2
InChI KeyREJHVSOVQBJEBF-OWOJBTEDSA-N
SMILESNC1=CC=C(\C=C\C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Acid-base back titration≥94.0 to ≤106.0%
Appearance (Color)Pale yellow to pale orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only