Tetraethyleneglycol monomethyl ether, 98%
Tetraethyleneglycol monomethyl ether, 98%
Tetraethyleneglycol monomethyl ether, 98%
Alfa Aesar

Tetraethyleneglycol monomethyl ether, 98%

CAS: 23783-42-8 | C9H20O5 | 208.25 g/mol
Have Questions?
Catalog NumberQuantity
ALF-A17021-22100 g
Catalog number ALF-A17021-22
HSN 29094900
Price (INR)
62,976.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
62,976.00
Each
Chemical Identifiers
CAS23783-42-8
IUPAC Name2,5,8,11-tetraoxatridecan-13-ol
Molecular FormulaC9H20O5
InChI KeyZNYRFEPBTVGZDN-UHFFFAOYSA-N
SMILESCOCCOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4435-1.4475 @ 20?C
FormLiquid

A very small fraction (less than 5%) may be used for other purposes, such as asphalt anti-stripping applications or as a chemical intermediate in other industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A very small fraction (less than 5%) may be used for other purposes, such as asphalt anti-stripping applications or as a chemical intermediate in other industrial processes.

Solubility
Water solubility 999,999c.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only