2-Chlorobutane, 98+%
2-Chlorobutane, 98+%
2-Chlorobutane, 98+%
Alfa Aesar

2-Chlorobutane, 98+%

CAS: 78-86-4 | C4H9Cl | 92.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1000 mL
Catalog number ALF-B21586-AK
HSN 29031990
Price (INR)
3,711.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (INR)
3,711.00
Each
Chemical Identifiers
CAS78-86-4
IUPAC Name2-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyBSPCSKHALVHRSR-UHFFFAOYNA-N
SMILESCCC(C)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.3945-1.3985 @ 20?C
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

Solubility
Immiscible with water.

Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Saunders, W. H.; Gronert, S. The Competition between Elimination Pathways in the Reactions of a Wide Variety of Bases with 2-Fluoro- and 2-Chlorobutane in the Gas Phase. J. Org. Chem. 2015, 80 (21), 10787-10793.
  2. Benito, J.; Guerrero, H.; Artigas, H.; López, M. C.; Lafuente, C. Refractive indices and static permittivities of systems containing n-hexane or n-heptane and isomeric chlorobutanes. J. Chem. Thermodyn. 2015, 86, 162-167.