4-Methyloctanoic acid, 98%
4-Methyloctanoic acid, 98%
4-Methyloctanoic acid, 98%
Alfa Aesar

4-Methyloctanoic acid, 98%

CAS: 54947-74-9 | C9H17O2 | 157.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF-B22616-06
HSN 29159090
Price (INR)
3,687.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
3,687.00
Each
Chemical Identifiers
CAS54947-74-9
IUPAC Name(4R)-4-methyloctanoate
Molecular FormulaC9H17O2
InChI KeyLEGGANXCVQPIAI-MRVPVSSYSA-M
SMILESCCCC[C@@H](C)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4315-1.4355 @ 20?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only