Rubrene, 97%
Rubrene, 97%
Rubrene, 97%
Alfa Aesar

Rubrene, 97%

CAS: 517-51-1 | C42H28 | 532.686 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALF-H53507-06
HSN 29029090
Price (INR)
44,716.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
44,716.00
Each
Chemical Identifiers
CAS517-51-1
IUPAC Name5,6,11,12-tetraphenyltetracene
Molecular FormulaC42H28
InChI KeyYYMBJDOZVAITBP-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=C2C(C3=CC=CC=C3)=C3C=CC=CC3=C(C3=CC=CC=C3)C2=C(C2=CC=CC=C2)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormCrystalline solid
Assay (HPLC)>97%
Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

Solubility
Soluble in hot toluene. Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cho, E. H.; Song, W. G.; Park, C. J.; Kim, J.; Kim, S.; Joo, J. Enhancement of photoresponsive electrical characteristics of multilayer MoS2 transistors using rubrene patches. Nano Res. 2015, 8 (3), 790-800.
  2. Sutton, C.; Marshall, M. S.; Sherrill, C. D.; Risko, C.; Bredas, J. L. Rubrene: The interplay between intramolecular and intermolecular interactions determines the planarization of its tetracene core in the solid state. J. Am. Chem. Soc. 2015, 137 (27), 8775-8782.