4-Nitrobenzenediazonium tetrafluoroborate, 97%
4-Nitrobenzenediazonium tetrafluoroborate, 97%
4-Nitrobenzenediazonium tetrafluoroborate, 97%
Alfa Aesar

4-Nitrobenzenediazonium tetrafluoroborate, 97%

CAS: 456-27-9 | C6H4BF4N3O2 | 236.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALF-H55315-14
HSN 29270090
Price (INR)
54,212.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
54,212.00
Each
Chemical Identifiers
CAS456-27-9
IUPAC Name4-nitrobenzene-1-diazonium; tetrafluoroboranuide
Molecular FormulaC6H4BF4N3O2
InChI KeyAFULQCYCQAHYIP-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4'-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

Solubility
Soluble in acetonitrile and dimethyl sulfoxide.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.
RUO – Research Use Only

General References:

  1. Brisset, F.; Vieillard, J.; Berton, B.; Grognet, S. M.; Poc, C. D.; Derf, F. L. Surface functionalization of cyclic olefin copolymer with aryldiazonium salts: A covalent grafting method. Appl. Surf. Sci. 2015, 329, 337-346.
  2. Mooste, M.; Kibena, E.; Kozlova, J.; Marandi, M.; Matisen, L.; Niilisk, A.; Sammelselg, V.; Tammeveski, K. Electrografting and morphological studies of chemical vapour deposition grown graphene sheets modified by electroreduction of aryldiazonium salts. Electrochim. Acta 2015, 161, 195-204.