Triphenyl borate, 97%
Triphenyl borate, 97%
Triphenyl borate, 97%
Alfa Aesar

Triphenyl borate, 97%

CAS: 1095-03-0 | C18H15BO3 | 290.125 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF-H55686-06
HSN 29319090
Price (INR)
4,002.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
4,002.00
Each
Chemical Identifiers
CAS1095-03-0
IUPAC Nametriphenyl borate
Molecular FormulaC18H15BO3
InChI KeyMDCWDBMBZLORER-UHFFFAOYSA-N
SMILESO(B(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥96.0%
FormPowder
Appearance (Color)Pale yellow to yellow
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

Solubility
Soluble in chloroform, methanol.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.
RUO – Research Use Only

General References:

  1. T Colclough.; W Gerrard.;MF Lappert. The preparation and properties of triphenyl borate and the phenoxyboron chlorides. Journal of the Chemical Society . 195555 (1), 907-911.
  2. MO Abdalla.; A Ludwick.; T Mitchell. Boron-modified phenolic resins for high performance applications. Polymer. 200344 (24), 7353-7359.