Adenosine-5'-triphosphate disodium salt hydrate, 98%
Adenosine-5'-triphosphate disodium salt hydrate, 98%
Adenosine-5'-triphosphate disodium salt hydrate, 98%
Alfa Aesar

Adenosine-5'-triphosphate disodium salt hydrate, 98%

CAS: 34369-07-8 | C10H14N5Na2O13P3 | 551.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
10 g
Catalog number ALF-J61125-14
HSN 29349900
Price (INR)
5,852.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
5,852.00
Each
Chemical Identifiers
CAS34369-07-8
IUPAC Namedisodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC10H14N5Na2O13P3
InChI KeyTTWYZDPBDWHJOR-WCYUCLFNNA-L
SMILES[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms to structure
Water Content (Karl Fischer Titration)≤10%
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay from Supplier's CofA≥98.0% (HPLC)
View more
Adenosine-5'-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gordon. Extracellular ATP: effects, sources and fate. Biochem.J. 1986, 233,(2), 309-19.
  2. Guilbert et al. Dependence of P2-nucleotide receptor agonist-mediated endothelium-indepdent relaxation on ectonucleotidase acticity and A2A-receptors in rat portal vein. Br.J.Pharm. 1998, 123,(2), 1732-35.