Sodium pyruvate, Cell Culture Grade
Sodium pyruvate, Cell Culture Grade
Sodium pyruvate, Cell Culture Grade
Alfa Aesar

Sodium pyruvate, Cell Culture Grade

CAS: 113-24-6 | C3H3NaO3 | 110.044 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number ALF-J61840-18
HSN 29183090
Price (INR)
23,462.00
Each
Quantity:
50 g
Request bulk or custom format
Price (INR)
23,462.00
Each
Chemical Identifiers
CAS113-24-6
IUPAC Namesodium 2-oxopropanoate
Molecular FormulaC3H3NaO3
InChI KeyDAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES[Na+].CC(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥99.0% (dried basis)
Loss on Drying≤2.0%
Heavy Metals≤10 ppm (as Pb)
pH5.0-7.0
Appearance (Color)White to cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only