Acrylamide (30%) Solutions
Acrylamide (30%) Solutions
Acrylamide (30%) Solutions
Alfa Aesar

Acrylamide (30%) Solutions

CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALF-J62100-AP
HSN 29241900
Price (INR)
13,194.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (INR)
13,194.00
Each
Chemical Identifiers
CAS79-06-1
IUPAC Nameprop-2-enamide
Molecular FormulaC3H5NO
InChI KeyHRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILESNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
CommentComposition: Acrylamide, 30% concentration
FormLiquid
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from Acids, Oxidizing agents, Iron and iron salts., Copper, Brass, Free radical initiators.
RUO – Research Use Only

General References:

  1. Molloy, M.P., et al. Extraction of membrane proteins by differential solubilization for separation using two-dimensional gel electrophoresis. Electrophoresis. 1998, 19, (5), 837-44.
  2. Rabilloud, T. Two-dimensional gel electrophoresis in proteomics: old, old fashioned, but it still climbs up the mountains. Proteomics. 2002, 2, (1), 3-10.