Sulindac Sulfide
Sulindac Sulfide
Sulindac Sulfide
Alfa Aesar

Sulindac Sulfide

CAS: 32004-67-4 | C20H17FO2S, C20H17FO2S | 340.41 g/mol
Have Questions?
Catalog NumberQuantity
ALF-J62104-MC100 mg
Catalog number ALF-J62104-MC
HSN 29309099
Price (INR)
77,508.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (INR)
77,508.00
Each
Specifications
Quantity100 mg
DescriptionYellow
For Use With (Application)Active metabolite of sulindac; inhibits cyclooxygenase-1
Recommended StorageAmbient temperatures
CAS32004-67-4
View more
It is also shown to inhibit growth and induce apoptosis in human prostate cancer cells through a COX-1 and COX-2 independent mechanism. Sulindac sulfide is an analgesic that has antiproliferative and apoptotic effects. It inhibits the expression and activity of cyclooxygenase-2 in human colon cancer cells, and reduces tumor burden in adenomatous polyposis patients. Human endothelial cells, HMEC-1 were treated with Sulindac sulfide and the effect on cell survival was studies by MTT assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only