MOPS-SDS running buffer (20X)
Alfa Aesar

MOPS-SDS running buffer (20X)

MOPS-SDS running buffer (20X) is a buffer frequently used for separating medium- to large-sized proteins on a gel.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALF-J62847-AP
HSN 98020000
Price (INR)
21,778.00
Each
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMOPS-SDS running buffer
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
MOPS SDS Running Buffer is recommended for separating medium- to large-sized proteins. MOPS-SDS Running Buffer is suitable for protein gel electrophoresis. Use of MOPS buffer allows proteins to run slower than when using MES buffer. This buffer is used in 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis - See more at:

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • MOPS-SDS running buffer is a zwitterionic, morpholinic buffer that allows the separation of medium- to large-sized proteins

Application

  • MOPS is used as a running buffer in electrophoresis. The use of MOPS buffer allows proteins to run slower than when MES buffer is used
  • MOPS-SDS running buffer, 20X concentrated, is formulated to be used for running proteins on an SDS gel
  • This buffer contains SDS detergent to maintain the denaturation of proteins and MOPS to buffer the pH
  • It is commonly used for cell culture media and protein purification in chromatography
  • MOPS is frequently applied in buffered culture media for bacteria, yeast, and mammalian cells
  • It is also applied in the bicinchoninic acid (BCA) assay
  • It is used in separating RNA in agarose gels
RUO – Research Use Only
MM Santoro.; DW Bolen. A test of the linear extrapolation of unfolding free energy changes over an extended denaturant concentration range. Biochemistry. 1992, 31, (20), 4901-4907.