Tween 20 washing buffer, 1% in PBS (10X)
Alfa Aesar

Tween 20 washing buffer, 1% in PBS (10X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2 L
Catalog number ALF-J63314-AP
HSN 98020000
Price (INR)
8,060.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (INR)
8,060.00
Each
Specifications
Chemical Name or MaterialBuffer
Recommended StorageKeep cold
Concentration1% in PBS (10X)
Physical FormViscous Liquid
Quantity500 mL
View more
Tween-20 is a non-ionic surfactant that is useful for solubilizing membrane proteins during isolation. Tween-20 is also added to buffers for immunohistochemistry. Tween-20 may also help to prevent non-specific antibody binding and function as a blocking agent in immunoblotting assays. It has been used in preextraction of membranes to remove peripheral proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only