7-Hydroxycoumarin, 98%
7-Hydroxycoumarin, 98%
7-Hydroxycoumarin, 98%
Alfa Aesar

7-Hydroxycoumarin, 98%

CAS: 93-35-6 | C9H6O3 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF-L04082-14
HSN 29322090
Price (INR)
4,938.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
4,938.00
Each
Chemical Identifiers
CAS93-35-6
IUPAC Name7-hydroxy-2H-chromen-2-one
Molecular FormulaC9H6O3
InChI KeyORHBXUUXSCNDEV-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC(=O)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Melting Point228-236?C
Appearance (Color)White to cream to yellow to orange to brown
Assay (HPLC)≥97.5%

7-Hydroxycoumarin is used as an intracellular pH sensitive fluorescent indicator & bloodbrain barrier probe. It is used as a sunscreen agent, and an optical brightener for textiles. It has also been used as a gain medium for dye lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Hydroxycoumarin is used as an intracellular pH sensitive fluorescent indicator & bloodbrain barrier probe. It is used as a sunscreen agent, and an optical brightener for textiles. It has also been used as a gain medium for dye lasers.

Solubility
Soluble in water (slightly at 20°C), 95% ethanol (50 mg/ml), chloroform, acetone, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Michaela Walshe; Joshua Howarth; Mary T Kelly; Richard O'Kennedy; Malcolm R Smyth. The preparation of a molecular imprinted polymer to 7- hydroxycoumarin and its use as a solid-phase extraction material.Journal of Pharmaceutical and Biomedical Analysis.1997, 16 319-325.
  2. M. E. Marshall; J. L. Mohler; K. Edmonds; B. Williams; K. Butler; M. Ryles; L. Weiss; D. Urban; A. Bueschen; M. Markiewicz; G. Cloud. An updated review of the clinical development of coumarin (1,2-benzopyrone) and 7-hydroxycoumarin.Journal of Cancer Research and Clinical Oncology.1994, 120 S39-S42.
  3. Fluorescent indicator.